• USA Home
  • Search Results

  • 2acetylpyrazine122122204725211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Acetylpyrazine

1 Products
Synonym:   Acetylpyrazine
Image of 2-Acetylpyrazine
CAS Number:
Empirical Formula (Hill Notation):
 C6H6N2O
Molecular Weight:
 122.12
 Council of Europe no.:
 2286
 EC Number:
 FEMA Number:
 3126
 Flavis number:
 14.032
 MDL number:
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More