• USA Home
  • Search Results

  • 2acetylpyridine12114112262911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Acetylpyridine

3 Products
Synonym:   Methyl 2-pyridyl ketone
Image of 2-Acetylpyridine
CAS Number:
Empirical Formula (Hill Notation):
 C7H7NO
Molecular Weight:
 121.14
 Beilstein/REAXYS Number:
 107759
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
  Anal. Standard Form
  Application(s)
  Assay
  Physical Form
Show More