• USA Home
  • Search Results

  • 2acetylpyrrole10913107283911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Acetylpyrrole

1 Products
Synonym:   Methyl 2-pyrrolyl ketone
Image of 2-Acetylpyrrole
CAS Number:
Empirical Formula (Hill Notation):
 C6H7NO
Molecular Weight:
 109.13
 Beilstein/REAXYS Number:
 1882
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Special Grade
Show More