• USA Home
  • Search Results

  • 2amino46dichloropyrimidine163995605311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Amino-4,6-dichloropyrimidine

1 Products
Image of 2-<WBR>Amino-<WBR>4,6-<WBR>dichloropyrimidine
CAS Number:
Empirical Formula (Hill Notation):
 C4H3Cl2N3
Molecular Weight:
 163.99
 Beilstein/REAXYS Number:
 118454
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More