• USA Home
  • Search Results

  • 2amino4fluorobenzoicacid1551344632211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Amino-4-fluorobenzoic acid

1 Products
Synonym:   4-Fluoroanthranilic acid
Image of 2-<WBR>Amino-<WBR>4-<WBR>fluorobenzoic acid
CAS Number:
Linear Formula:
 H2NC6H3(F)CO2H
Molecular Weight:
 155.13
 Beilstein/REAXYS Number:
 2803665
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More