• USA Home
  • Search Results

  • 2amino5methylbenzoicacid15116294178811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Amino-5-methylbenzoic acid

1 Products
Synonym:   5-Methylanthranilic acid
Image of 2-<WBR>Amino-<WBR>5-<WBR>methylbenzoic acid
CAS Number:
Linear Formula:
 H2NC6H3(CH3)CO2H
Molecular Weight:
 151.16
 Beilstein/REAXYS Number:
 1101527
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More