• USA Home
  • Search Results

  • 2amino5nitrobenzoicacid1821361679511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Amino-5-nitrobenzoic acid

1 Products
Synonym:   5-Nitroanthranilic acid
Image of 2-<WBR>Amino-<WBR>5-<WBR>nitrobenzoic acid
CAS Number:
Linear Formula:
 O2NC6H3(NH2)CO2H
Molecular Weight:
 182.13
 Beilstein/REAXYS Number:
 646219
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More