• USA Home
  • Search Results

  • 2amino5nitrothiazole1451412166411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Amino-5-nitrothiazole

1 Products
Image of 2-<WBR>Amino-<WBR>5-<WBR>nitrothiazole
CAS Number:
Empirical Formula (Hill Notation):
 C3H3N3O2S
Molecular Weight:
 145.14
 Beilstein/REAXYS Number:
 126797
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More