• USA Home
  • Search Results

  • 2aminobenzimidazole1331593432711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Aminobenzimidazole

2 Products
Synonym:   2-Benzimidazolamine
Image of 2-Aminobenzimidazole
CAS Number:
Empirical Formula (Hill Notation):
 C7H7N3
Molecular Weight:
 133.15
 Beilstein/REAXYS Number:
 116525
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 32151902
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More