• USA Home
  • Search Results

  • 2aminoflubendazole255258205013311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Aminoflubendazole

1 Products
Synonym:   (2-Amino-1H-benzoimidazol-5-yl)-(4-fluoro-phenyl)-methanone, 2-Amino-5-(4-fluorobenzoyl)-1H-benzimidazole, Hydrolyzed flubendazole
Image of 2-Aminoflubendazole
CAS Number:
Empirical Formula (Hill Notation):
 C14H10FN3O
Molecular Weight:
 255.25
 Beilstein/REAXYS Number:
 8849496
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More