• USA Home
  • Search Results

  • 2aminopurine1351345206211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Aminopurine

1 Products
Synonym:   2-AP
Image of 2-Aminopurine
CAS Number:
Empirical Formula (Hill Notation):
 C5H5N5
Molecular Weight:
 135.13
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
A3509 2-Aminopurine, ≥99%,