• USA Home
  • Search Results

  • 2bromo132benzodioxaborole198815190185011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromo-1,3,2-benzodioxaborole

1 Products
Synonym:   B-Bromocatechol-borane
Image of 2-<WBR>Bromo-<WBR>1,3,2-<WBR>benzodioxaborole
CAS Number:
Empirical Formula (Hill Notation):
 C6H4BBrO2
Molecular Weight:
 198.81
 Beilstein/REAXYS Number:
 2445073
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More