• USA Home
  • Search Results

  • 2bromo1chloropropanesolution15744301795611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromo-1-chloropropane solution

2 Products
CAS Number:
Empirical Formula (Hill Notation):
 C3H6BrCl
Molecular Weight:
 157.44
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Agency/Method
  Anal. Standard Form
  Special Grade
  Package quantity
  Feature
  Concentration
Show More