• USA Home
  • Search Results

  • 2bromo2methylpropane1370250719711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromo-2-methylpropane

2 Products
Synonym:   2-methyl-2-bromopropane, tert-Butyl bromide
Image of 2-<WBR>Bromo-<WBR>2-<WBR>methylpropane
CAS Number:
Linear Formula:
 (CH3)3CBr
Molecular Weight:
 137.02
 Beilstein/REAXYS Number:
 1730892
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
Show More