• USA Home
  • Search Results

  • 2bromo2nitro13propanediol199995251711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromo-2-nitro-1,3-propanediol

1 Products
Synonym:   BNPD, BNPK, Bronopol
Image of 2-<WBR>Bromo-<WBR>2-<WBR>nitro-<WBR>1,3-<WBR>propanediol
CAS Number:
Linear Formula:
 HOCH2C(Br)(NO2)CH2OH
Molecular Weight:
 199.99
 Beilstein/REAXYS Number:
 1705868
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More