• USA Home
  • Search Results

  • 2bromo4fluorobenzoicacid21901100641311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromo-4-fluorobenzoic acid

1 Products
Image of 2-<WBR>Bromo-<WBR>4-<WBR>fluorobenzoic acid
CAS Number:
Linear Formula:
 BrC6H3(F)CO2H
Molecular Weight:
 219.01
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More