• USA Home
  • Search Results

  • 2bromo4fluorotoluene18902142253311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromo-4-fluorotoluene

1 Products
Image of 2-<WBR>Bromo-<WBR>4-<WBR>fluorotoluene
CAS Number:
Linear Formula:
 CH3C6H3(Br)F
Molecular Weight:
 189.02
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More