• USA Home
  • Search Results

  • 2bromobenzylalcohol187031898254211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromobenzyl alcohol

1 Products
Image of 2-Bromobenzyl alcohol
CAS Number:
Linear Formula:
 BrC6H4CH2OH
Molecular Weight:
 187.03
 Beilstein/REAXYS Number:
 2243418
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More