• USA Home
  • Search Results

  • 2bromobutane137027876211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromobutane

2 Products
Synonym:   sec-Butyl bromide
Image of 2-Bromobutane
CAS Number:
Linear Formula:
 CH3CH2CHBrCH3
Molecular Weight:
 137.02
 Beilstein/REAXYS Number:
 505949
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More