• USA Home
  • Search Results

  • 2bromoethylaminehydrobromide20489257647811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromoethylamine hydrobromide

2 Products
Synonym:   2-Aminoethyl bromide hydrobromide
Image of 2-Bromoethylamine hydrobromide
CAS Number:
Linear Formula:
 BrCH2CH2NH2 · HBr
Molecular Weight:
 204.89
 Beilstein Registry Number:
 3607605
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More