• USA Home
  • Search Results

  • 2bromohexanoicacid1950561605711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromohexanoic acid

1 Products
Synonym:   2-Bromocaproic acid
Image of 2-Bromohexanoic acid
CAS Number:
Linear Formula:
 CH3(CH2)3CHBrCOOH
Molecular Weight:
 195.05
 Beilstein/REAXYS Number:
 1721762
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More

Product #
Description
242837 2-Bromohexanoic acid, 97%,