• USA Home
  • Search Results

  • 2bromomethylnaphthalene2210993926411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-(Bromomethyl)naphthalene

1 Products
Image of 2-<WBR>(Bromomethyl)<WBR>naphthalene
CAS Number:
Linear Formula:
 C10H7CH2Br
Molecular Weight:
 221.09
 Beilstein/REAXYS Number:
 636546
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More