• USA Home
  • Search Results

  • 2bromoquinoline20805200543811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Bromoquinoline

2 Products
Image of 2-Bromoquinoline
CAS Number:
Empirical Formula (Hill Notation):
 C9H6BrN
Molecular Weight:
 208.05
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More