• USA Home
  • Search Results

  • 2butanol74127892211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Butanol

8 Products
Synonym:   sec-Butyl alcohol
Image of 2-Butanol
CAS Number:
Linear Formula:
 CH3CH2CH(OH)CH3
Molecular Weight:
 74.12
 Beilstein/REAXYS Number:
 773649
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Refractive Index
  Anal. Standard Form
  pH Value
  Vapor pressure
  Transition temperature
  Solubility
  Melting Point
  Explosion limit
  Density
  Application(s)
  Potency
  Autoignition temp
  Special Grade
  Physical Form
Show More