• USA Home
  • Search Results

  • 2butanone72117893311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Butanone

9 Products
Synonym:   Ethyl methyl ketone, MEK, Methyl ethyl ketone
Image of 2-Butanone
CAS Number:
Linear Formula:
 C2H5COCH3
Molecular Weight:
 72.11
 Beilstein/REAXYS Number:
 741880
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Restriction
  Refractive Index
  Document Links
  Allergen
  Organoleptic property
  Vapor pressure
  UV-Absorption Condition
  Transition temperature
  Application(s)
  Solubility
  Purity Grade
  Melting Point
  Ice Shipping
  Special Grade
  Transmittance
  Color
  Agency/Method
  UV-Absorption Values
  Assay
Show More