• USA Home
  • Search Results

  • 2butanoneoxime87129629711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Butanone oxime

1 Products
Synonym:   Methyl ethyl ketoxime
Image of 2-Butanone oxime
CAS Number:
Linear Formula:
 C2H5C(=NOH)CH3
Molecular Weight:
 87.12
 Beilstein/REAXYS Number:
 1698241
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
332828 2-Butanone oxime, 99%,