• USA Home
  • Search Results

  • 2butyn1ol700976401211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Butyn-1-ol

1 Products
Image of 2-Butyn-1-ol
CAS Number:
Linear Formula:
 CH3C≡CCH2OH
Molecular Weight:
 70.09
 Beilstein/REAXYS Number:
 1733676
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
244341 2-Butyn-1-ol, 98%,