• USA Home
  • Search Results

  • 2chloro132benzodioxaphosphorin4one20253538199711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Chloro-1,3,2-benzodioxaphosphorin-4-one

1 Products
Synonym:   SalPCl, Salicyl chlorophosphite
Image of 2-<WBR>Chloro-<WBR>1,3,2-<WBR>benzodioxaphosphorin-<WBR>4-<WBR>one
CAS Number:
Empirical Formula (Hill Notation):
 C7H4ClO3P
Molecular Weight:
 202.53
 Beilstein/REAXYS Number:
 1311951
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
Show More