• USA Home
  • Search Results

  • 2chloro2methylpropane925750720011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Chloro-2-methylpropane

3 Products
Synonym:   tert-Butyl chloride
Image of 2-<WBR>Chloro-<WBR>2-<WBR>methylpropane
CAS Number:
Linear Formula:
 (CH3)3CCl
Molecular Weight:
 92.57
 Beilstein/REAXYS Number:
 1730872
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
Show More