• USA Home
  • Search Results

  • 2chloro4fluorophenol14655199641411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Chloro-4-fluorophenol

1 Products
Image of 2-<WBR>Chloro-<WBR>4-<WBR>fluorophenol
CAS Number:
Linear Formula:
 ClC6H3(F)OH
Molecular Weight:
 146.55
 Beilstein/REAXYS Number:
 1861277
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More