• USA Home
  • Search Results

  • 2chloro4nitrophenol1735561908911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Chloro-4-nitrophenol

1 Products
Image of 2-<WBR>Chloro-<WBR>4-<WBR>nitrophenol
CAS Number:
Linear Formula:
 ClC6H3(NO2)OH
Molecular Weight:
 173.55
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More