• USA Home
  • Search Results

  • 2chloro6fluorophenylacetonitrile169587527955911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Chloro-6-fluorophenylacetonitrile

1 Products
Synonym:   2-Chloro-6-fluorobenzyl cyanide
Image of 2-<WBR>Chloro-<WBR>6-<WBR>fluorophenylacetonitrile
CAS Number:
Linear Formula:
 ClC6H3(F)CH2CN
Molecular Weight:
 169.58
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More