• USA Home
  • Search Results

  • 2chloroethylbenzene1406162224211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(2-Chloroethyl)benzene

1 Products
Synonym:   2-Phenylethyl chloride
Image of (2-<WBR>Chloroethyl)<WBR>benzene
CAS Number:
Linear Formula:
 C6H5CH2CH2Cl
Molecular Weight:
 140.61
 Beilstein/REAXYS Number:
 385969
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More