• USA Home
  • Search Results

  • 2chlorophenothiazine233729239711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Chlorophenothiazine

1 Products
Synonym:   2-Chlorothiodiphenylamine
Image of 2-<WBR>Chlorophenothiazine
CAS Number:
Empirical Formula (Hill Notation):
 C12H8ClNS
Molecular Weight:
 233.72
 Beilstein/REAXYS Number:
 176587
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More