• USA Home
  • Search Results

  • 2chlorothiophenol14462632003211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Chlorothiophenol

1 Products
Synonym:   2-Chlorobenzenethiol
Image of 2-Chlorothiophenol
CAS Number:
Linear Formula:
 ClC6H4SH
Molecular Weight:
 144.62
 Beilstein/REAXYS Number:
 1209665
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
247146 2-Chlorothiophenol, 99%,