• USA Home
  • Search Results

  • 2chlorotritylchloride313224207468011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Chlorotrityl chloride

3 Products
Synonym:   Chloro(2-chlorophenyl)diphenylmethane
Image of 2-Chlorotrityl chloride
CAS Number:
Empirical Formula (Hill Notation):
 C19H14Cl2
Molecular Weight:
 313.22
 Beilstein/REAXYS Number:
 1983914
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Melting Point
Show More