• USA Home
  • Search Results

  • 2cis4transabscisicacid264321437545211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-cis,4-trans-Abscisic acid

1 Products
Synonym:   (±)-Abscisic acid, (2Z,4E)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-2,4-pentadienoic acid, ABA, Dormin
Image of 2-<I>cis</I>,4-<I>trans</I>-Abscisic acid
CAS Number:
Empirical Formula (Hill Notation):
 C15H20O4
Molecular Weight:
 264.32
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More