• USA Home
  • Search Results

  • 2cyclohexen1one961393068711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Cyclohexen-1-one

1 Products
Image of 2-Cyclohexen-1-one
CAS Number:
Linear Formula:
 C6H8(=O)
Molecular Weight:
 96.13
 Beilstein/REAXYS Number:
 1280477
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More