• USA Home
  • Search Results

  • 2cyclohexylethanol12821444279911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Cyclohexylethanol

2 Products
Image of 2-Cyclohexylethanol
CAS Number:
Linear Formula:
 C₈H₁₆O
Molecular Weight:
 128.21 g/mol
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More