• USA Home
  • Search Results

  • 2decanone1562769354911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Decanone

4 Products
Synonym:   Methyl octyl ketone
Image of 2-Decanone
CAS Number:
Linear Formula:
 CH3(CH2)7COCH3
Molecular Weight:
 156.27
 Beilstein/REAXYS Number:
 1747463
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Document Links
  Allergen
  Application(s)
  Biological Source
Show More