• USA Home
  • Search Results

  • 2deoxyadenosinemonohydrate269261637393611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2′-Deoxyadenosine monohydrate

2 Products
Synonym:   9-(2-Deoxy-β-D-ribofuranosyl)adenine, Adenine deoxyriboside
Image of 2′-Deoxyadenosine monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C10H13N5O3 · H2O
Molecular Weight:
 269.26
 Beilstein/REAXYS Number:
 91015
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More