• USA Home
  • Search Results

  • 2deoxydglucose1641615417611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Deoxy-D-glucose

3 Products
Synonym:   2-Deoxy-D-arabinohexose, 2-Deoxyglucose
Image of 2-Deoxy-<SC>D</SC>-glucose
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O5
Molecular Weight:
 164.16
 Beilstein/REAXYS Number:
 1723331
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Color
  Physical Form
  Titertest
  Solubility
  Biological Source
Show More