• USA Home
  • Search Results

  • 2deoxyguanosine5triphosphatetrisodiumsaltsolution573139391941611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2′-Deoxyguanosine 5′-triphosphate trisodium salt solution

2 Products
Synonym:   dGTP-Na3
Image of 2′-Deoxyguanosine 5′-triphosphate trisodium salt solution
CAS Number:
Empirical Formula (Hill Notation):
 C10H13N5Na3O13P3
Molecular Weight:
 573.13
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More