• USA Home
  • Search Results

  • 2dicyclohexylphosphinomethyl13bis26diisopropylphenyl3imidazoliumiodide72680126015475311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-[(Dicyclohexylphosphino)methyl]-1,3-bis(2,6-diisopropylphenyl)-3-imidazolium iodide

2 Products
Synonym:   2-((Dicyclohexylphosphino)methyl)-1,3-bis(2,6-diisopropylphenyl)-1H-imidazol-3-ium iodide
Image of 2-<WBR>[(Dicyclohexylphosphino)<WBR>methyl]<WBR>-<WBR>1,3-<WBR>bis(2,6-<WBR>diisopropylphenyl)<WBR>-<WBR>3-<WBR>imidazolium iodide
CAS Number:
Empirical Formula (Hill Notation):
 C40H60IN2P
Molecular Weight:
 726.80
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Melting Point
Show More