• USA Home
  • Search Results

  • 2diethylaminoethanethiolhydrochloride16972194252511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Diethylaminoethanethiol hydrochloride

1 Products
Synonym:   2-Diethylaminoethyl mercaptan hydrochloride
Image of 2-<WBR>Diethylaminoethanethiol hydrochloride
CAS Number:
Linear Formula:
 (C2H5)2NCH2CH2SH · HCl
Molecular Weight:
 169.72
 Beilstein/REAXYS Number:
 3548721
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More