• USA Home
  • Search Results

  • 2diisopropylaminoethylchloridehydrochloride20015426168111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-(Diisopropylamino)-ethylchloride hydrochloride

1 Products
Synonym:   2-(Diisopropylamino)-ethylchloride hydrochloride, N-(2-Chloroethyl)-diisopropylamine hydrochloride
Image of 2-<WBR>(Diisopropylamino)<WBR>-<WBR>ethylchloride hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C8H19Cl2N
Molecular Weight:
 200.15
 EC Index Number
 224-238-1
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More