• USA Home
  • Search Results

  • 2ethoxyethylacetate1321611115911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Ethoxyethyl acetate

3 Products
Synonym:   1-Acetoxy-2-ethoxyethane, Cellosolve® acetate, Ethylene glycol monoethyl ether acetate
Image of 2-Ethoxyethyl acetate
CAS Number:
Linear Formula:
 CH3COOCH2CH2OC2H5
Molecular Weight:
 132.16
 Beilstein/REAXYS Number:
 1748677
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Solubility
  Physical Form
  Application(s)
Show More