• USA Home
  • Search Results

  • 2ethoxyphenol138169471311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Ethoxyphenol

1 Products
Synonym:   Guaethol, Pyrocatechol monoethyl ether
Image of 2-Ethoxyphenol
CAS Number:
Linear Formula:
 C2H5OC6H4OH
Molecular Weight:
 138.16
 Beilstein/REAXYS Number:
 1100453
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
250910 2-Ethoxyphenol, 98%,