• USA Home
  • Search Results

  • 2ethyl1hexanol1302310476711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Ethyl-1-hexanol

5 Products
Synonym:   Isooctyl alcohol
Image of 2-Ethyl-1-hexanol
CAS Number:
Linear Formula:
 CH3(CH2)3CH(C2H5)CH2OH
Molecular Weight:
 130.23
 Beilstein/REAXYS Number:
 1719280
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Application(s)
  Potency
  Special Grade
Show More