• USA Home
  • Search Results

  • 2ethyl2phenylmalonamidemonohydrate224268086690611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Ethyl-2-phenylmalonamide monohydrate

1 Products
Synonym:   PEMA
Image of 2-<WBR>Ethyl-<WBR>2-<WBR>phenylmalonamide monohydrate
CAS Number:
Linear Formula:
 C6H5C(C2H5)(CONH2)2·H2O
Molecular Weight:
 224.26
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More